Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286834.1 | NP_001273763.1 | 4, 5 | 618 | |
NM_001286837.1 | NP_001273766.1 | 4, 5 | 856 | ||
NM_002936.4 | NP_002927.2 | 4, 5 | 618 | ||
GenBank mRNA | AF039652.1 | - | 4, 5 | 588 | |
AF048994.1 | - | 4, 5 | 534 | ||
AF048995.1 | - | 4, 5 | 568 | ||
AJ224117.1 | - | 4, 5 | 507 | ||
BC002973.1 | - | 4, 5 | 577 | 3537074 | |
CR541820.1 | - | 4, 5 | 507 |