Gene Transcripts

Gene Symbol
RNASE4
Entrez Gene ID
6038
Gene Name
ribonuclease A family member 4
Gene Aliases
RAB1, RNS4
Chromosome Location
Chr. 14: 20684213 - 20700602 on Build GRCh38
UniGene ID
Hs.283749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282192.1 NP_001269121.1 3 1310
NM_001282193.1 NP_001269122.1 2 1144
NM_002937.4 NP_002928.1 2 1206
NM_194431.2 NP_919412.1 2 758
GenBank mRNA AK092422.1 - 2 1206
BC015520.1 - 2 815 3905439
BX247985.1 - 2 996
BX248010.1 - 3 864
D37931.1 - 1 670

View Full Product Details