Gene Transcripts

Gene Symbol
PI4K2A
Entrez Gene ID
55361
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
PI4KII, PIK42A
Chromosome Location
Chr. 10: 97640686 - 97676434 on Build GRCh38
UniGene ID
Hs.25300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018425.3 NP_060895.1 1 469
GenBank mRNA AJ303098.1 - 1 469
BC003167.1 - 1 441 2905670
BT007330.1 - 1 412

View Full Product Details