Gene Transcripts

Gene Symbol
PITHD1
Entrez Gene ID
57095
Gene Name
PITH domain containing 1
Gene Aliases
C1orf128, HT014, TXNL1CL
Chromosome Location
Chr. 1: 23778386 - 23788232 on Build GRCh38
UniGene ID
Hs.31819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020362.4 NP_065095.2 5 622
XM_011541804.1 XP_011540106.1 4 547
GenBank mRNA AF218024.1 - 5 603
AF221595.1 - 5 614
AF271784.1 - 5 618
AK296662.1 - 4 547
AK310330.1 - 5 569
AK313005.1 - 5 590
BC017208.2 - 5 583 3943360

View Full Product Details