Gene Transcripts

Gene Symbol
TP53AIP1
Entrez Gene ID
63970
Gene Name
tumor protein p53 regulated apoptosis inducing protein 1
Gene Aliases
P53AIP1
Chromosome Location
Chr. 11: 128934732 - 128944233 on Build GRCh38
UniGene ID
Hs.160953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251964.1 NP_001238893.1 2 2057
GenBank mRNA AB045832.1 - 2 1585

View Full Product Details