Gene Transcripts
- Gene Symbol
- INTS3
- Entrez Gene ID
- 65123
- Gene Name
- integrator complex subunit 3
- Gene Aliases
- C1orf193, C1orf60, INT3, SOSS-A, SOSSA
- Chromosome Location
- Chr. 1: 153728050 - 153774808 on Build GRCh38
- UniGene ID
- Hs.438723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001324475.1 |
NP_001311404.1
|
2, 3 |
382 |
|
NM_023015.4 |
NP_075391.3
|
1, 2 |
723 |
|
XM_005245459.2 |
XP_005245516.1
|
1, 2 |
740 |
|
GenBank mRNA |
AK290025.1 |
- |
2, 3 |
376 |
|
AL832133.1 |
- |
1, 2 |
107 |
|
BC105092.1 |
- |
1, 2 |
186 |
8144095 |
BC105094.1 |
- |
1, 2 |
186 |
8144097 |
BC116458.1 |
- |
1, 2 |
718 |
5752962 |
BX640786.1 |
- |
1, 2 |
709 |
|
BX640950.1 |
- |
1, 2 |
724 |
|
CR627233.1 |
- |
1, 2 |
723 |
|
CR749212.1 |
- |
1, 2 |
717 |
|
CR749376.1 |
- |
1, 2 |
724 |
|
View Full Product Details