Gene Transcripts
- Gene Symbol
- PAAF1
- Entrez Gene ID
- 80227
- Gene Name
- proteasomal ATPase associated factor 1
- Gene Aliases
- PAAF, Rpn14, WDR71
- Chromosome Location
- Chr. 11: 73876699 - 73927736 on Build GRCh38
- UniGene ID
- Hs.525017
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267803.1 |
NP_001254732.1
|
7, 8 |
650 |
|
NM_001267804.1 |
NP_001254733.1
|
7, 8 |
641 |
|
NM_001267805.1 |
NP_001254734.1
|
7, 8 |
637 |
|
NM_001267806.1 |
NP_001254735.1
|
6, 7 |
524 |
|
NM_025155.2 |
NP_079431.1
|
6, 7 |
581 |
|
XM_005274308.2 |
XP_005274365.2
|
4, 5 |
1173 |
|
XM_005274309.1 |
XP_005274366.1
|
5, 6 |
440 |
|
XM_017018357.1 |
XP_016873846.1
|
7, 8 |
569 |
|
XM_017018358.1 |
XP_016873847.1
|
7, 8 |
574 |
|
XM_017018359.1 |
XP_016873848.1
|
4, 5 |
1132 |
|
GenBank mRNA |
AF087895.1 |
- |
7, 8 |
575 |
|
AK021910.1 |
- |
6, 7 |
562 |
|
AK222501.1 |
- |
6, 7 |
562 |
|
AK225520.1 |
- |
6, 7 |
549 |
|
AK298258.1 |
- |
6, 7 |
524 |
|
AK309133.1 |
- |
4, 5 |
788 |
|
AK316394.1 |
- |
6, 7 |
795 |
|
BC006142.2 |
- |
6, 7 |
534 |
3937356 |
BC021541.1 |
- |
6, 7 |
545 |
3942680 |
BC028628.1 |
- |
7, 8 |
615 |
5263875 |
CD243380.1 |
- |
7, 8 |
627 |
|
View Full Product Details