Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032315.2 | NP_115691.1 | 5 | 692 | |
XM_005263503.3 | XP_005263560.1 | 5 | 361 | ||
XM_011542296.1 | XP_011540598.1 | 5 | 479 | ||
GenBank mRNA | AF495714.1 | - | 5 | 660 | |
AJ880283.1 | - | 5 | 678 | ||
BC004991.1 | - | 5 | 662 | 2905957 | |
BC073135.1 | - | 5 | 678 | 6172562 |