Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 2 327
NM_032793.4 NP_116182.2 2 327
NR_109896.1 2 327
XM_005271285.1 XP_005271342.1 2 291
XM_011542312.2 XP_011540614.1 2 294
GenBank mRNA AB073383.1 - 2 291
AF289609.1 - 2 283
AK074186.1 - 2 232
AK075183.1 - 2 294
AK093223.1 - 2 289
AK291540.1 - 2 289
AL834184.1 - 2 281
AY358636.1 - 2 308
BC011587.1 - 2 301 4646430
BC092414.1 - 2 269 30387073
BP321361.1 - 2 327
DC297497.1 - 2 327

View Full Product Details