Gene Transcripts
- Gene Symbol
- CLRN3
- Entrez Gene ID
- 119467
- Gene Name
- clarin 3
- Gene Aliases
- TMEM12, USH3AL1
- Chromosome Location
- Chr. 10: 127877841 - 127892947 on Build GRCh38
- UniGene ID
- Hs.242014
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029478.1 |
- |
1 |
265 |
|
View Full Product Details