Gene Transcripts

Gene Symbol
CLRN3
Entrez Gene ID
119467
Gene Name
clarin 3
Gene Aliases
TMEM12, USH3AL1
Chromosome Location
Chr. 10: 127877841 - 127892947 on Build GRCh38
UniGene ID
Hs.242014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152311.3 NP_689524.1 3 935
XM_011539274.2 XP_011537576.1 2 754
GenBank mRNA BC029478.1 - 3 935
BX640823.1 - 2 1215

View Full Product Details