Gene Transcripts
- Gene Symbol
- CCDC17
- Entrez Gene ID
- 149483
- Gene Name
- coiled-coil domain containing 17
- Gene Aliases
- Chromosome Location
- Chr. 1: 45620044 - 45624787 on Build GRCh38
- UniGene ID
- Hs.18912
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057646.1 |
- |
11 |
1853 |
|
View Full Product Details