Gene Transcripts

Gene Symbol
CNIH2
Entrez Gene ID
254263
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnil
Chromosome Location
Chr. 11: 66278201 - 66284214 on Build GRCh38
UniGene ID
Hs.437072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182553.2 NP_872359.1 6 1275
NR_073078.1 6 1301
NR_073079.1 6 1271
GenBank mRNA BC047953.1 - 6 1251

View Full Product Details