Gene Transcripts
- Gene Symbol
- ANKRD45
- Entrez Gene ID
- 339416
- Gene Name
- ankyrin repeat domain 45
- Gene Aliases
- CT117
- Chromosome Location
- Chr. 1: 173608336 - 173705430 on Build GRCh38
- UniGene ID
- Hs.130054
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198493.2 |
NP_940895.1
|
2, 3 |
397 |
|
XM_011509466.2 |
XP_011507768.1
|
3, 4 |
844 |
|
XM_011509467.1 |
XP_011507769.1
|
4, 5 |
1368 |
|
XM_011509468.1 |
XP_011507770.1
|
3, 4 |
1336 |
|
XM_011509470.1 |
XP_011507772.1
|
2, 3 |
1052 |
|
XM_011509471.2 |
XP_011507773.1
|
3, 4 |
2036 |
|
XM_017001123.1 |
XP_016856612.1
|
4, 5 |
706 |
|
XM_017001124.1 |
XP_016856613.1
|
3, 4 |
2036 |
|
GenBank mRNA |
AK127170.1 |
- |
2, 3 |
397 |
|
BC126353.1 |
- |
2, 3 |
397 |
8992069 |
BC126355.1 |
- |
2, 3 |
397 |
8992071 |
View Full Product Details