Gene Transcripts
- Gene Symbol
- PHLDB3
- Entrez Gene ID
- 653583
- Gene Name
- pleckstrin homology like domain family B member 3
- Gene Aliases
- Chromosome Location
- Chr. 19: 43474341 - 43504916 on Build GRCh38
- UniGene ID
- Hs.631581
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097512.1 |
- |
6 |
1420 |
|
BC070094.1 |
- |
6 |
1713 |
4374205 |
View Full Product Details