Gene Transcripts

Gene Symbol
SKI
Entrez Gene ID
6497
Gene Name
SKI proto-oncogene
Gene Aliases
SGS, SKV
Chromosome Location
Chr. 1: 2228695 - 2310213 on Build GRCh38
UniGene ID
Hs.656507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003036.3 NP_003027.1 1 1004
XM_005244775.3 XP_005244832.1 1 974
GenBank mRNA X15218.1 - 1 1004

View Full Product Details