Gene Transcripts

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
Chromosome Location
Chr. 9: 129612225 - 129622275 on Build GRCh38
UniGene ID
Hs.124223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199350.3 NP_955382.3 3 885
XM_011518655.1 XP_011516957.1 5 1440
XM_011518656.1 XP_011516958.1 4 950
XM_011518657.2 XP_011516959.1 4 681
XM_011518658.2 XP_011516960.1 4 681
XM_011518659.1 XP_011516961.1 4 681
XM_011518660.1 XP_011516962.1 4 681
XM_011518663.1 XP_011516965.1 4 681
XM_017014708.1 XP_016870197.1 4 681
GenBank mRNA AK093122.1 - 3 885
JF442034.1 - 3 356

View Full Product Details