Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134367.3 | NP_001127839.2 | 3 | 451 | |
NM_001134368.3 | NP_001127840.1 | 3 | 455 | ||
NM_003043.5 | NP_003034.2 | 3 | 455 | ||
NR_103507.2 | 3 | 455 | |||
XM_006713307.2 | XP_006713370.1 | 2 | 259 | ||
XM_011534030.2 | XP_011532332.1 | 2 | 212 | ||
XM_011534031.2 | XP_011532333.1 | 3 | 454 | ||
GenBank mRNA | AB209172.1 | - | 3 | 381 | |
AK303649.1 | - | 3 | 455 | ||
AK307284.1 | - | 3 | 410 | ||
AK309534.1 | - | 3 | 406 | ||
BC006252.2 | - | 3 | 385 | ||
BC111489.1 | - | 3 | 411 | ||
BC137128.1 | - | 3 | 402 | 9021125 | |
BC137129.1 | - | 3 | 398 | 9021126 | |
BM149397.1 | - | 3 | 425 | ||
DC359099.1 | - | 3 | 410 | ||
U09220.1 | - | 1 | 130 | ||
U16120.1 | - | 4 | 506 | ||
Z18956.1 | - | 1 | 149 |