Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014177.2 | NP_054896.2 | 6 | 1146 | |
XM_005266689.3 | XP_005266746.1 | 6 | 1147 | ||
GenBank mRNA | AF161503.1 | - | 6 | 1056 | |
AK289923.1 | - | 6 | 1144 | ||
BC000892.1 | - | 6 | 1106 | 3463640 | |
BG820086.1 | - | 1 | 125 |