Gene Transcripts

Gene Symbol
PDS5A
Entrez Gene ID
23244
Gene Name
PDS5 cohesin associated factor A
Gene Aliases
PIG54, SCC-112, SCC112
Chromosome Location
Chr. 4: 39822863 - 39977956 on Build GRCh38
UniGene ID
Hs.331431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100399.1 NP_001093869.1 25 3453
XM_011513672.2 XP_011511974.1 25 3066
XM_017007928.1 XP_016863417.1 25 3028
GenBank mRNA AB014548.1 - 13 1457
AB621803.1 - 25 3467
AF294791.1 - 25 3024
BC009650.1 - 1 38
BC114218.1 - 25 3149 5579832
BC126225.1 - 24 2863 8991941
CR749425.1 - 24 2822

View Full Product Details