Gene Transcripts

Gene Symbol
CNRIP1
Entrez Gene ID
25927
Gene Name
cannabinoid receptor interacting protein 1
Gene Aliases
C2orf32, CRIP1
Chromosome Location
Chr. 2: 68284171 - 68320051 on Build GRCh38
UniGene ID
Hs.212885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015463.2 NP_056278.1 3 1881
XM_005264249.1 XP_005264306.1 4 1576
XM_006711984.1 XP_006712047.1 4 1344
GenBank mRNA AK092542.1 - 1 2522
AL110235.1 - 3 1229
BC011535.1 - 3 1541 3870150
BC035125.1 - 1 4053

View Full Product Details