Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018008.3 | NP_060478.3 | 5 | 1517 | |
GenBank mRNA | AF064845.1 | - | 5 | 1357 | |
AF332890.1 | - | 4 | 1369 | ||
AK290464.1 | - | 5 | 1517 | ||
BC022464.1 | - | 5 | 1348 | 4794121 |