Gene Transcripts

Gene Symbol
CAMKV
Entrez Gene ID
79012
Gene Name
CaM kinase like vesicle associated
Gene Aliases
1G5, VACAMKL
Chromosome Location
Chr. 3: 49857981 - 49870222 on Build GRCh38
UniGene ID
Hs.145156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320147.1 NP_001307076.1 12 2433
NM_024046.4 NP_076951.2 11 2526
GenBank mRNA AK074856.1 - 11 2240
AK074899.1 - 12 2117
AL136697.1 - 10 2125
AL833943.1 - 9 2584
AL834461.1 - 1 315
BC000497.2 - 11 2195 2820734
BC001921.1 - 11 2195 2820734
BC005828.2 - 11 2195 2820734
BC017363.2 - 11 2197 3940018
BC019256.1 - 11 2190 3502894
BC033374.1 - 1 54
BX647074.1 - 10 2368

View Full Product Details