Gene Transcripts

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
Chromosome Location
Chr. 19: 13882348 - 13906125 on Build GRCh38
UniGene ID
Hs.143288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024323.3 NP_077299.3 6 769
XM_006722885.3 XP_006722948.2 6 1142
XM_011528286.2 XP_011526588.1 5 931
XM_011528287.2 XP_011526589.1 5 591
XM_017027300.1 XP_016882789.1 5 929
XM_017027301.1 XP_016882790.1 5 793
GenBank mRNA AK097431.1 - 6 769
BC012945.1 - 2 345
BC119719.1 - 5 737 40117748

View Full Product Details