Gene Transcripts

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
Chromosome Location
Chr. 3: 101724098 - 101770562 on Build GRCh38
UniGene ID
Hs.444135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303401.1 NP_001290330.1 4 465
NM_024548.3 NP_078824.2 4 465
XM_011513127.1 XP_011511429.1 4 422
GenBank mRNA AK294784.1 - 4 403
AL833269.1 - 4 415
BC041085.1 - 4 408 5528397
DA520309.1 - 4 467

View Full Product Details