Gene Transcripts

Gene Symbol
C3orf36
Entrez Gene ID
80111
Gene Name
chromosome 3 open reading frame 36
Gene Aliases
Chromosome Location
Chr. 3: 133928145 - 133929812 on Build GRCh38
UniGene ID
Hs.287691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025041.2 NP_079317.2 1 1570
GenBank mRNA AK025826.1 - 1 1432
AK225383.1 - 1 1432
BC051274.1 - 1 1570
BC064502.1 - 1 1570
BC082257.1 - 1 1570
BC104161.1 - 1 749 40029559
BC104162.1 - 1 746 40029562

View Full Product Details