Gene Transcripts
- Gene Symbol
- RFT1
- Entrez Gene ID
- 91869
- Gene Name
- RFT1 homolog
- Gene Aliases
- CDG1N
- Chromosome Location
- Chr. 3: 53071151 - 53130469 on Build GRCh38
- UniGene ID
- Hs.631910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052859.3 |
NP_443091.1
|
7, 8 |
837 |
|
XM_005265537.4 |
XP_005265594.1
|
7, 8 |
850 |
|
XM_006713384.3 |
XP_006713447.1
|
7, 8 |
848 |
|
XM_011534214.2 |
XP_011532516.1
|
7, 8 |
852 |
|
XM_011534215.2 |
XP_011532517.1
|
7, 8 |
852 |
|
XM_011534216.2 |
XP_011532518.1
|
7, 8 |
935 |
|
XM_017007460.1 |
XP_016862949.1
|
7, 8 |
849 |
|
XM_017007461.1 |
XP_016862950.1
|
8, 9 |
1021 |
|
GenBank mRNA |
AJ318099.1 |
- |
7, 8 |
821 |
|
BC043595.1 |
- |
7, 8 |
830 |
5769824 |
View Full Product Details