Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080862.2 | NP_543138.1 | 3 | 1864 | |
GenBank mRNA | AF403029.1 | - | 3 | 1374 | |
AK056147.1 | - | 3 | 709 | ||
AK056367.1 | - | 3 | 1335 | ||
BC008324.1 | - | 3 | 1374 | 3501702 |