Gene Transcripts

Gene Symbol
KLHDC7B
Entrez Gene ID
113730
Gene Name
kelch domain containing 7B
Gene Aliases
Chromosome Location
Chr. 22: 50548033 - 50551023 on Build GRCh38
UniGene ID
Hs.137007
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138433.3 NP_612442.2 1 1279
GenBank mRNA BC009980.2 - 1 1279

View Full Product Details