Gene Transcripts

Gene Symbol
TTC21A
Entrez Gene ID
199223
Gene Name
tetratricopeptide repeat domain 21A
Gene Aliases
IFT139A, STI2
Chromosome Location
Chr. 3: 39107661 - 39139009 on Build GRCh38
UniGene ID
Hs.443935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105513.2 NP_001098983.2 16, 17 2381
NM_145755.2 NP_665698.2 17, 18 2525
XM_005264921.4 XP_005264978.1 17, 18 2454
XM_005264922.4 XP_005264979.1 17, 18 2454
XM_005264923.3 XP_005264980.1 17, 18 2454
XM_005264924.4 XP_005264981.1 16, 17 2343
XM_005264925.4 XP_005264982.1 16, 17 2331
XM_005264927.4 XP_005264984.1 13, 14 1908
XM_006713011.3 XP_006713074.1 17, 18 2454
XM_011533447.2 XP_011531749.1 16, 17 2355
GenBank mRNA AJ487015.1 - 17, 18 2487
AK093313.1 - 7, 8 1065
AK302408.1 - 16, 17 2364
AK302465.1 - 16, 17 2235
BC129948.1 - 17, 18 2436 40126472

View Full Product Details