Gene Transcripts

Gene Symbol
HORMAD2
Entrez Gene ID
150280
Gene Name
HORMA domain containing 2
Gene Aliases
CT46.2
Chromosome Location
Chr. 22: 30080069 - 30207425 on Build GRCh38
UniGene ID
Hs.120391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011529914.2 XP_011528216.1 9 934
XM_011529917.2 XP_011528219.1 9 793
XM_011529918.1 XP_011528220.1 9 934
XM_017028621.1 XP_016884110.1 9 815
XM_017028622.1 XP_016884111.1 9 734
XM_017028624.1 XP_016884113.1 8 897
XM_017028625.1 XP_016884114.1 4 285
XM_017028626.1 XP_016884115.1 2 228
GenBank mRNA AK098703.1 - 9 759
BC030013.1 - 9 544 4823215
BI459310.1 - 8 502
CR456341.1 - 8 521

View Full Product Details