Gene Transcripts
- Gene Symbol
- TANGO2
- Entrez Gene ID
- 128989
- Gene Name
- transport and golgi organization 2 homolog
- Gene Aliases
- C22orf25, MECRCN
- Chromosome Location
- Chr. 22: 20017000 - 20067164 on Build GRCh38
- UniGene ID
- Hs.474233
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041339.1 |
- |
5, 6 |
563 |
|
BI544307.1 |
- |
5, 6 |
563 |
|
View Full Product Details