Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152945.3 | NP_694453.2 | 1 | 229 | |
XM_005246287.4 | XP_005246344.1 | 1 | 239 | ||
XM_017003320.1 | XP_016858809.1 | 1 | 243 | ||
XM_017003321.1 | XP_016858810.1 | 1 | 244 | ||
XM_017003322.1 | XP_016858811.1 | 1 | 238 | ||
GenBank mRNA | AB036991.1 | - | 1 | 151 | |
AF526533.1 | - | 1 | 229 | ||
AK126575.1 | - | 1 | 189 | ||
BC066549.1 | - | 1 | 198 | 5748065 | |
BI763677.1 | - | 1 | 229 |