Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173566.2 | NP_775837.2 | 5 | 5963 | |
XM_006724219.3 | XP_006724282.1 | 5 | 6523 | ||
XM_006724220.3 | XP_006724283.1 | 5 | 5925 | ||
XM_006724221.2 | XP_006724284.1 | 4 | 5479 | ||
XM_011530080.2 | XP_011528382.1 | 6 | 6731 | ||
XM_011530081.2 | XP_011528383.1 | 5 | 6607 | ||
XM_017028736.1 | XP_016884225.1 | 5 | 7683 | ||
XM_017028737.1 | XP_016884226.1 | 4 | 7121 | ||
GenBank mRNA | AK130944.1 | - | 2 | 1777 | |
BC040859.1 | - | 2 | 677 | 5535580 | |
BX647272.1 | - | 2 | 266 | ||
CT841517.1 | - | 4 | 5795 |