Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
Chromosome Location
Chr. 21: 42062959 - 42142996 on Build GRCh38
UniGene ID
Hs.242520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004416.2 NP_001004416.2 20 3562
NM_001199527.1 NP_001186456.1 19 4126
NM_001199528.2 NP_001186457.2 20 3742
NM_173568.3 NP_775839.3 19 3946
XM_011529797.1 XP_011528099.1 17 3554
XM_017028506.1 XP_016883995.1 19 3941
XM_017028507.1 XP_016883996.1 19 6870
XM_017028508.1 XP_016883997.1 9 1725
GenBank mRNA AB051810.1 - 19 4126
AB051811.1 - 20 3742
AB096973.1 - 1 26
AK093654.1 - 5 804
AK124223.1 - 6 1306
AY771619.1 - 19 3946
AY771620.1 - 20 3562

View Full Product Details