Gene Transcripts

Gene Symbol
CMC1
Entrez Gene ID
152100
Gene Name
C-X9-C motif containing 1
Gene Aliases
C3orf68
Chromosome Location
Chr. 3: 28241588 - 28322874 on Build GRCh38
UniGene ID
Hs.444724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182523.1 NP_872329.1 4 483
XM_011533402.2 XP_011531704.1 6 660
XM_017005760.1 XP_016861249.1 5 442
GenBank mRNA AF086095.1 - 2 645
BC052644.1 - 4 483 6163781
BX648671.1 - 4 510
CR749370.1 - 3 3618

View Full Product Details