Gene Transcripts

Gene Symbol
WDR53
Entrez Gene ID
348793
Gene Name
WD repeat domain 53
Gene Aliases
Chromosome Location
Chr. 3: 196554188 - 196568542 on Build GRCh38
UniGene ID
Hs.385865
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182627.1 NP_872433.1 3 700
GenBank mRNA AK314105.1 - 3 709
BC054030.1 - 3 700 6145598
EF011620.1 - 3 700

View Full Product Details