Gene Transcripts

Gene Symbol
CCDC22
Entrez Gene ID
28952
Gene Name
coiled-coil domain containing 22
Gene Aliases
CXorf37, JM1, RTSC2
Chromosome Location
Chr. X: 49235467 - 49250526 on Build GRCh38
UniGene ID
Hs.26333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014008.4 NP_054727.1 7 1059
XM_005272599.3 XP_005272656.1 7 988
GenBank mRNA AJ005890.1 - 7 975
AK291976.1 - 7 1020
AK296911.1 - 7 1030
AK298264.1 - 7 1054
AK316375.1 - 7 1005
BC000972.2 - 7 1006 3449051
BC011675.2 - 7 890 4299954

View Full Product Details