Gene Transcripts
- Gene Symbol
- SNTA1
- Entrez Gene ID
- 6640
- Gene Name
- syntrophin alpha 1
- Gene Aliases
- LQT12, SNT1, TACIP1, dJ1187J4.5
- Chromosome Location
- Chr. 20: 33407957 - 33443892 on Build GRCh38
- UniGene ID
- Hs.31121
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003098.2 |
NP_003089.1
|
2, 3 |
775 |
|
XM_005260517.1 |
XP_005260574.1
|
2, 3 |
736 |
|
XM_011529007.1 |
XP_011527309.1
|
2, 3 |
736 |
|
XM_011529008.1 |
XP_011527310.1
|
2, 3 |
736 |
|
GenBank mRNA |
AK095942.1 |
- |
2, 3 |
335 |
|
AK291994.1 |
- |
2, 3 |
646 |
|
AK301800.1 |
- |
2, 3 |
506 |
|
BC026215.2 |
- |
2, 3 |
615 |
4413197 |
S81737.1 |
- |
2, 3 |
635 |
|
U40571.1 |
- |
2, 3 |
540 |
|
View Full Product Details