Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304529.1 | NP_001291458.1 | 7 | 1874 | |
NM_001304530.1 | NP_001291459.1 | 7 | 2083 | ||
NM_001304532.1 | NP_001291461.1 | 8 | 2129 | ||
NM_018246.2 | NP_060716.2 | 9 | 2181 | ||
NR_130761.1 | 7 | 2031 | |||
NR_130762.1 | 7 | 2025 | |||
XM_011544570.2 | XP_011542872.1 | 8 | 2128 | ||
XM_011544571.2 | XP_011542873.1 | 7 | 2007 | ||
GenBank mRNA | AK001715.1 | - | 8 | 2129 | |
BC065295.1 | - | 9 | 2123 | 6139413 |