Gene Transcripts

Gene Symbol
PLCXD1
Entrez Gene ID
55344
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 1
Gene Aliases
LL0XNC01-136G2.1
Chromosome Location
Chr. Y: 276324 - 303356 on Build GRCh38
UniGene ID
Hs.522568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018390.3 NP_060860.1 7 4970
NR_028057.1 9 1753
XM_006724866.2 XP_006724929.1 7 4722
GenBank mRNA AK002185.1 - 10 1755
AK091006.1 - 1 2151
AK097346.1 - 1 1408
BC005028.1 - 8 1530 3953383
BC064136.1 - 3 592

View Full Product Details