Gene Transcripts

Gene Symbol
ARMCX6
Entrez Gene ID
54470
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
GASP10
Chromosome Location
Chr. X: 101615118 - 101618001 on Build GRCh38
UniGene ID
Hs.83530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009584.1 NP_001009584.1 2 1319
NM_001184768.1 NP_001171697.1 4 1489
NM_019007.3 NP_061880.2 3 1401
NR_033669.1 4 490
NR_033670.1 5 578
XM_011530976.2 XP_011529278.1 4 1455
GenBank mRNA AK000818.1 - 1 983
AK097018.1 - 1 1793
AW955663.1 - 5 490
BC007677.2 - 3 1401 3609980
BX423052.2 - 4 491
L20773.1 - 1 1119

View Full Product Details