Gene Transcripts

Gene Symbol
CENPU
Entrez Gene ID
79682
Gene Name
centromere protein U
Gene Aliases
CENP50, CENPU50, KLIP1, MLF1IP, PBIP1
Chromosome Location
Chr. 4: 184694065 - 184734132 on Build GRCh38
UniGene ID
Hs.575032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024629.3 NP_078905.2 6 476
NR_104593.1 6 476
XM_005263218.4 XP_005263275.2 6 511
GenBank mRNA AF469667.1 - 6 479
AF516710.1 - 7 893
AK027121.1 - 4 286
AK292297.1 - 6 422
BC067802.1 - 6 416
BC131556.1 - 6 476 40108113
BC141854.1 - 6 474 8860370
DQ907910.1 - 6 406
DQ907911.1 - 6 406

View Full Product Details