Gene Transcripts

Gene Symbol
ABHD18
Entrez Gene ID
80167
Gene Name
abhydrolase domain containing 18
Gene Aliases
C4orf29
Chromosome Location
Chr. 4: 127965256 - 128039711 on Build GRCh38
UniGene ID
Hs.143995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319305.1 NP_001306234.1 11 1357
NM_001319306.1 NP_001306235.1 10 1312
NM_001319307.1 NP_001306236.1 11 1375
XM_005263238.4 XP_005263295.1 11 1408
XM_006714318.3 XP_006714381.1 12 1453
XM_006714320.3 XP_006714383.1 12 1241
XM_006714322.3 XP_006714385.1 8 791
XM_011532272.2 XP_011530574.1 13 1520
XM_011532273.2 XP_011530575.1 12 1475
XM_011532274.2 XP_011530576.1 11 1351
XM_011532276.1 XP_011530578.1 11 1351
XM_011532278.2 XP_011530580.1 10 1214
XM_011532279.2 XP_011530581.1 5 727
XM_017008631.1 XP_016864120.1 10 1306
XM_017008632.1 XP_016864121.1 5 641
GenBank mRNA AK024759.1 - 10 1124
BC034253.1 - 10 1190 4339231
BC111400.1 - 10 1292
BC128142.1 - 10 1062 40112521
BC128143.1 - 10 1005 40112522

View Full Product Details