Gene Transcripts
- Gene Symbol
- TTC29
- Entrez Gene ID
- 83894
- Gene Name
- tetratricopeptide repeat domain 29
- Gene Aliases
- NYD-SP14, TBPP2A
- Chromosome Location
- Chr. 4: 146706638 - 146945886 on Build GRCh38
- UniGene ID
- Hs.378893
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300761.2 |
NP_001287690.1
|
11, 12 |
1442 |
|
NM_001317806.1 |
NP_001304735.1
|
10, 11 |
1341 |
|
NM_031956.3 |
NP_114162.2
|
10, 11 |
1341 |
|
NR_133922.1 |
|
11, 12 |
1495 |
|
XM_005263270.1 |
XP_005263327.1
|
11, 12 |
1424 |
|
XM_006714335.1 |
XP_006714398.1
|
11, 12 |
1424 |
|
XM_006714336.1 |
XP_006714399.1
|
11, 12 |
1424 |
|
XM_006714339.2 |
XP_006714402.1
|
10, 11 |
1238 |
|
GenBank mRNA |
AF345910.1 |
- |
10, 11 |
1292 |
|
AK093145.1 |
- |
11, 12 |
1388 |
|
AK225725.1 |
- |
11, 12 |
1442 |
|
AK301807.1 |
- |
10, 11 |
1267 |
|
AK314066.1 |
- |
10, 11 |
1337 |
|
BC024193.2 |
- |
10, 11 |
1282 |
4825866 |
EF432564.1 |
- |
8, 9 |
1110 |
|
View Full Product Details