Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317889.1 | NP_001304818.1 | 3 | 165 | |
NM_001317891.1 | NP_001304820.1 | 2 | 99 | ||
NM_032026.3 | NP_114415.1 | 3 | 165 | ||
NR_027427.1 | 3 | 165 | |||
XM_006716666.3 | XP_006716729.1 | 3 | 185 | ||
XM_006716667.3 | XP_006716730.1 | 3 | 154 | ||
XM_006716668.3 | XP_006716731.1 | 3 | 167 | ||
XM_011517331.2 | XP_011515633.1 | 2 | 81 | ||
XM_017013895.1 | XP_016869384.1 | 3 | 175 | ||
XM_017013896.1 | XP_016869385.1 | 3 | 154 | ||
XM_017013897.1 | XP_016869386.1 | 2 | 75 | ||
XM_017013899.1 | XP_016869388.1 | 3 | 161 | ||
GenBank mRNA | AF212250.1 | - | 3 | 148 | |
AK312204.1 | - | 3 | 161 | ||
BC064964.1 | - | 3 | 149 | 6141834 | |
BG529611.1 | - | 2 | 90 | ||
BX332564.2 | - | 3 | 147 |