Gene Transcripts

Gene Symbol
PRRC1
Entrez Gene ID
133619
Gene Name
proline rich coiled-coil 1
Gene Aliases
Chromosome Location
Chr. 5: 127517609 - 127555089 on Build GRCh38
UniGene ID
Hs.483259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ427339.1 - 8 1245
AJ441110.3 - 8 1245
AJ515158.1 - 8 1245
AJ515428.1 - 8 1245
AJ515429.1 - 8 1245

View Full Product Details