Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003128.2 | NP_003119.2 | 4 | 629 | |
NM_178313.2 | NP_842565.2 | 3 | 726 | ||
XM_005264517.2 | XP_005264574.1 | 4 | 538 | ||
XM_005264518.1 | XP_005264575.1 | 3 | 1384 | ||
XM_006712087.2 | XP_006712150.1 | 4 | 502 | ||
XM_017004779.1 | XP_016860268.1 | 4 | 645 | ||
XM_017004780.1 | XP_016860269.1 | 4 | 599 | ||
XM_017004781.1 | XP_016860270.1 | 4 | 600 | ||
GenBank mRNA | AB209748.1 | - | 4 | 507 | |
AB371586.1 | - | 4 | 619 | ||
AF327441.1 | - | 3 | 726 | ||
BC032061.1 | - | 4 | 474 | ||
BC137282.1 | - | 3 | 636 | 9021279 | |
BC137283.1 | - | 3 | 636 | 9021280 | |
EU794662.1 | - | 4 | 629 | ||
JQ599728.1 | - | 3 | 380 |