Gene Transcripts

Gene Symbol
CLDN23
Entrez Gene ID
137075
Gene Name
claudin 23
Gene Aliases
CLDNL, hCG1646163
Chromosome Location
Chr. 8: 8702156 - 8704107 on Build GRCh38
UniGene ID
Hs.183617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194284.2 NP_919260.2 1 1798
GenBank mRNA AK123547.1 - 2 1423

View Full Product Details