Gene Transcripts

Gene Symbol
FAM221A
Entrez Gene ID
340277
Gene Name
family with sequence similarity 221 member A
Gene Aliases
C7orf46
Chromosome Location
Chr. 7: 23680114 - 23703249 on Build GRCh38
UniGene ID
Hs.592178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127364.2 NP_001120836.1 2 300
NM_199136.4 NP_954587.2 2 300
XM_011515369.2 XP_011513671.1 2 295
XM_011515370.2 XP_011513672.1 2 295
XM_011515371.2 XP_011513673.1 2 295
XM_011515372.2 XP_011513674.1 2 440
XM_017012134.1 XP_016867623.1 2 295
XM_017012135.1 XP_016867624.1 2 440
GenBank mRNA AK127774.1 - 2 247
AL832127.1 - 2 236
BC042034.1 - 2 282
BC063130.1 - 2 214 30382529
BI549491.1 - 2 287
CA867596.1 - 2 261
HY056255.1 - 2 301

View Full Product Details